Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TRMU Full-length ORF (AAH80631.1, 1 a.a. - 222 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number AAH80631.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55687
Molecular Weight (g/mol) 51.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-965-831
View Documents
Abnova™
H00055687P01S
10 ug
Each for $499.00
Add to Cart
 
89-965-832
View Documents
Abnova™
H00055687P01L
25 ug
Each for $769.00
Add to Cart
 
Description

Description

This gene is a member of the trmU family. It encodes a mitochondria-specific tRNA-modifying enzyme that is required for the 2-thio modification of 5-taurinomethyl-2-thiouridine tRNA-Lys on the wobble position of the anticodon. [provided by RefSeq]

Sequence: MKKSLSRSTLRSPNGFSEIGLKLEMVSQDALRRTIFPLGGLTKEFVKKIAAENRLHHVLQKKESMGMCFIGKRNFEHFLLQYLQPRPGHFISIEDNKVLGTHKGWFLYTLGQRANIGGLREPWYVVEKDSVKGDVFVAPRTDHPALYRDLLRTSRVHWIAEEPPAALVRDKMMECHFRFRHQMALVCCVLQGGRVPGQREDPAAGAVCLHAPEGPAQSWDGH
Specifications

Specifications

AAH80631.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.6kDa
Glutathione Sepharose 4 Fast Flow
MKKSLSRSTLRSPNGFSEIGLKLEMVSQDALRRTIFPLGGLTKEFVKKIAAENRLHHVLQKKESMGMCFIGKRNFEHFLLQYLQPRPGHFISIEDNKVLGTHKGWFLYTLGQRANIGGLREPWYVVEKDSVKGDVFVAPRTDHPALYRDLLRTSRVHWIAEEPPAALVRDKMMECHFRFRHQMALVCCVLQGGRVPGQREDPAAGAVCLHAPEGPAQSWDGH
RUO
TRMU
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
55687
TRMU (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC99627/MTO2/MTU1/TRMT/TRMT1/TRNT1
TRMU
Recombinant
wheat germ expression system
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit