Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TUB Partial ORF (NP_003311, 439 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00007275Q01S

 View more versions of this product

Catalog No. 89-966-153


Add to cart

Description

Description

This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: KMSVIVPGMNMVHERVSIRPRNEHETLLARWQNKNTESIIELQNKTPVWNDDTQSYVLNFHGRVTQASVKNFQIIHGNDPDYIVMQFGRVAEDVFTMDYN
Specifications

Specifications

NP_003311
Liquid
7275
TUB (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
rd5
TUB
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KMSVIVPGMNMVHERVSIRPRNEHETLLARWQNKNTESIIELQNKTPVWNDDTQSYVLNFHGRVTQASVKNFQIIHGNDPDYIVMQFGRVAEDVFTMDYN
RUO
TUB
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit