Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TUBD1 Full-length ORF (AAH00258.1, 1 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name TUBD1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 71.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-966-193
View Documents
Abnova™
H00051174P01L
25 ug
Each for $709.50
Add to cart
 
89-966-192
View Documents
Abnova™
H00051174P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

Sequence: MSIVTVQLGQCGNQIGFEVFDALLSDSHSSQGLCSMRENEAYQASCKERFFSEEENGVPIARAVLVDMEPKVINQMLSKAAQSGQWKYGQHACFCQKQGSGNNWAYGYSVHGPRHEESIMNIIRKEVEKCDSFSGFFIIMSMAGGTGSGLGAFVTQNLEDQYSNSLKMNQIIWPYGTGEVIVQNYNSILTLSHLYRSSDALLLHENDAIHKICAKLMNIKQISFSDINQVLAHQLGSVFQPTYSAESSFHYRRNPLGIDRHVWPPLSGLPPLSKMSLNKDLHFNTSIANLVILRGKDVQSADVEGFKDPALYTSWLKPVNAFNVWKTQRAFSKYEKSAVLVSNSQFLVKPLDMIVGKAWNMFASKAYIHQYTKFGIEEEDFLDSFTSLEQVVASYCNL
Specifications

Specifications

TUBD1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
71.1kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit