Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TWEAK/TNFSF12 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $2020.00

Specifications

Antigen TWEAK/TNFSF12
Clone 173830
Concentration LYOPH
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications ELISA
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1090SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1090SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB1090100
GREENER_CHOICE
View Documents
R&D Systems
MAB1090100
100 μg
Each for $549.50
Only null left
Add to Cart
 
MAB1090500
GREENER_CHOICE
View Documents
R&D Systems
MAB1090500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

TWEAK/TNFSF12 Monoclonal specifically detects TWEAK/TNFSF12 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

TWEAK/TNFSF12
LYOPH
ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
TNFSF12
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
173830
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
O43508
8742
E. coli-derived recombinant human TWEAK/TNFSF12 Arg93-His249 Accession # O43508
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human TWEAK/TNFSF12 in ELISAs. In sandwich immunoassays, approximately 15% reactivity with recombinant mouse TWEAK is observed and no cross-reactivity with recombinant human (rh) BAFF, rhEDA-A2, rhLIGHT, rhFas Ligand, rhGITR Ligand, rhLT alpha 1 beta 2, rhLT alpha 2 beta 1, rhOX40 Ligand, rhTNF-alpha , rhTRANCE, rhTRAIL, rh4-1BB, or rhAPRIL is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.