Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human TWEAK/TNFSF12 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1090
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Polyclonal specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.Specifications
TWEAK/TNFSF12 | |
Polyclonal | |
Unconjugated | |
Q4ACW9 | |
TNFSF12 | |
E. coli-derived recombinant human TWEAK Arg93-His249 Accession # Q4ACW9 | |
100 μg | |
Primary | |
Detects human TWEAK in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) BAFF, rhFas Ligand, rhGITR Ligand, rhOX40 Ligand, rhTRAIL and less than 1% cross-reactivity with rhAPRIL, rhLIGHT, rhTNF-alpha , rhVEGI, and rhTRANCE is observed. | |
Human | |
IgG |
Western Blot, Neutralization, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 2-8 ug/mL, Immunocytochemistry 5-15 ug/mL | |
APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
Goat | |
Affinity Purified | |
RUO | |
8742 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction