Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Tyk2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3427SP
This item is not returnable.
View return policy
Description
Tyk2 Monoclonal specifically detects Tyk2 in Human samples. It is validated for Western Blot.Specifications
Tyk2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P29597 | |
TYK2 | |
E. coli-derived recombinant human Tyk2 Pro2-Leu51 Accession # P29597 | |
25 μg | |
Primary | |
Detects human Tyk2 in Western blots. | |
Human | |
Purified |
Western Blot | |
307214 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.2, JTK1, non-receptor tyrosine-protein kinase TYK2, tyrosine kinase 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7297 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction