Learn More
Bioworld HUMAN UBIQUITIN (RECOMBINANT)
Supplier:  Bioworld 220605081
Encompass_Preferred
Recombinant Human Ubiquitin (Ub) Source: E.Coli Background: Recombinant Human Ubiquitin (Ub) is a small protein composed of 76 amino acids and having a molecular mass of 8.6kDa. Ub is found only in eukaryotic organisms among which it...
Catalog No. 50-253-605