Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UBQLN3 Partial ORF (NP_059509, 401 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00050613Q01L

 View more versions of this product

Catalog No. 89-966-469


Add to cart

Description

Description

Summary: This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This gene is specifically expressed in the testis, and proposed to regulate cell-cycle progression during spermatogenesis. [provided by RefSeq]

Sequence: VAIKGRSSCPAFLRYPTENSTGQGGDQDGAGKSSTGHSTNLPDLVSGLGDSANRVPFAPLSFSPTAAIPGIPEPPWLPSPAYPRSLRPDGMNPAPQLQDE
Specifications

Specifications

NP_059509
Liquid
50613
UBQLN3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TUP-1
UBQLN3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VAIKGRSSCPAFLRYPTENSTGQGGDQDGAGKSSTGHSTNLPDLVSGLGDSANRVPFAPLSFSPTAAIPGIPEPPWLPSPAYPRSLRPDGMNPAPQLQDE
RUO
UBQLN3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit