Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UCHL5IP Full-length ORF (NP_059988.3, 1 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055559P01S

 View more versions of this product

Catalog No. 89-966-597


Add to cart

Description

Description

This gene encodes a protein identified by interaction with ubiquitin C-terminal hydrolase 37, which functions to edit polyubiquitin chains on ubiquitinated substrates. This protein is a subunit of the multisubunit augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. [provided by RefSeq]

Sequence: MGGARLGARNMAGQDAGCGRGGDDYSEDEGDSSVSRAAVEVFGKLKDLNCPFLEGLYITEPKTIQELLCSPSEYRLEILEWMCTRVWPSLQDRFSSLKGVPTEVKIQEMTKLGHELMLCAPDDQELLKGCACAQKQLHFMDQLLDTIRSLTIGCSSCSSLMEHFEDTREKNEALLGELFSSPHLQMLLNPECDPWPLDMQPLLNKQSDDWQWASASAKSEEEEKLAELARQLQESAAKLHALRTEYFAQHEQGAAAGAADISTLDQKLRLVTSDFHQLILAFLQVYDDELGECCQRPGPDLHPCGPIIQATHQNLTSYSQLLQVVMAVADTSAKAVETVKKQQGEQICWGGSSSVMSLATKMNELMEK
Specifications

Specifications

NP_059988.3
Liquid
55559
UCHL5IP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGGARLGARNMAGQDAGCGRGGDDYSEDEGDSSVSRAAVEVFGKLKDLNCPFLEGLYITEPKTIQELLCSPSEYRLEILEWMCTRVWPSLQDRFSSLKGVPTEVKIQEMTKLGHELMLCAPDDQELLKGCACAQKQLHFMDQLLDTIRSLTIGCSSCSSLMEHFEDTREKNEALLGELFSSPHLQMLLNPECDPWPLDMQPLLNKQSDDWQWASASAKSEEEEKLAELARQLQESAAKLHALRTEYFAQHEQGAAAGAADISTLDQKLRLVTSDFHQLILAFLQVYDDELGECCQRPGPDLHPCGPIIQATHQNLTSYSQLLQVVMAVADTSAKAVETVKKQQGEQICWGGSSSVMSLATKMNELMEK
RUO
UCHL5IP
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSXQ28ORF|UIP1
UCHL5IP
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit