Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UGT8 Partial ORF (NP_003351, 31 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00007368Q01S

 View more versions of this product

Catalog No. 89-966-587


Add to cart

Description

Description

Galactocerebrosides are abundant sphingolipids of the myelin membrane of the central nervous system and peripheral nervous system and are also present in small amounts in kidney. The key enzymatic step in the biosynthesis of galactocerebrosides consists of the transfer of galactose to ceramide catalyzed by UDP-galactose ceramide galactosyltransferase (CGT, EC 2.4.1.45). The enzyme encoded by the CGT gene is the first involved in complex lipid biosynthesis in the myelinating oligodendrocyte.[supplied by OMIM]

Sequence: FESHMYIFKTLASALHERGHHTVFLLSEGRDIAPSNHYSLQRYPGIFNSTTSDAFLQSKMRNIFSGRLTAIELFDILDHYTKNCDLMVGNHALIQGLKKEKFDLLLVDPN
Specifications

Specifications

NP_003351
Liquid
7368
UGT8 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CGT
UGT8
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FESHMYIFKTLASALHERGHHTVFLLSEGRDIAPSNHYSLQRYPGIFNSTTSDAFLQSKMRNIFSGRLTAIELFDILDHYTKNCDLMVGNHALIQGLKKEKFDLLLVDPN
RUO
UGT8
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit