Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UNC13B Partial ORF (NP_006368, 1482 a.a. - 1591 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010497Q01S

 View more versions of this product

Catalog No. 89-966-636


Add to cart

Description

Description

This gene is expressed in the kidney cortical epithelial cells and is upregulated by hyperglycemia. The encoded protein shares a high level of similarity to the rat homolog, and contains 3 C2 domains and a diacylglycerol-binding C1 domain. Hyperglycemia increases the levels of diacylglycerol, which has been shown to induce apoptosis in cells transfected with this gene and thus contribute to the renal cell complications of hyperglycemia. Studies in other species also indicate a role for this protein in the priming step of synaptic vesicle exocytosis. [provided by RefSeq]

Sequence: SKSNNWAPKYNETFHLLLGNEEGPESYELQICVKDYCFAREDRVLGLAVMPLRDVTAKGSCACWCPLGRKIHMDETGLTILRILSQRSNDEVAREFVKLKSESRSTEEGS
Specifications

Specifications

NP_006368
Liquid
10497
UNC13B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC133279|MGC133280|MUNC13|UNC13|Unc13h2|hmunc13
UNC13B
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SKSNNWAPKYNETFHLLLGNEEGPESYELQICVKDYCFAREDRVLGLAVMPLRDVTAKGSCACWCPLGRKIHMDETGLTILRILSQRSNDEVAREFVKLKSESRSTEEGS
RUO
UNC13B
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit