Learn More
Abnova™ Human UNC5C Partial ORF (AAH41156, 498 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH41156 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8633 |
Molecular Weight (g/mol) | 36.41kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-966-651
|
Abnova™
H00008633Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-966-650
|
Abnova™
H00008633Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin (Ig)-like domains and 2 type I thrombospondin motifs in the extracellular region. [provided by RefSeq]
Sequence: IKVYNTSGAVTPQDDLSEFTSKLSPQMTQSLLENEALSLKNQSLARQTDPSCTAFGSFNSLGGHLIVPNSGVSLLIPAGAIPQGRVYEMYVTVHRKETMRSpecifications
AAH41156 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UNC5H3 | |
UNC5C | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8633 | |
UNC5C (Human) Recombinant Protein (Q01) | |
IKVYNTSGAVTPQDDLSEFTSKLSPQMTQSLLENEALSLKNQSLARQTDPSCTAFGSFNSLGGHLIVPNSGVSLLIPAGAIPQGRVYEMYVTVHRKETMR | |
RUO | |
UNC5C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.