Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UNC5C Partial ORF (AAH41156, 498 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008633Q01S

 View more versions of this product

Catalog No. 89-966-650


Add to cart

Description

Description

This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin (Ig)-like domains and 2 type I thrombospondin motifs in the extracellular region. [provided by RefSeq]

Sequence: IKVYNTSGAVTPQDDLSEFTSKLSPQMTQSLLENEALSLKNQSLARQTDPSCTAFGSFNSLGGHLIVPNSGVSLLIPAGAIPQGRVYEMYVTVHRKETMR
Specifications

Specifications

AAH41156
Liquid
8633
UNC5C (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UNC5H3
UNC5C
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IKVYNTSGAVTPQDDLSEFTSKLSPQMTQSLLENEALSLKNQSLARQTDPSCTAFGSFNSLGGHLIVPNSGVSLLIPAGAIPQGRVYEMYVTVHRKETMR
RUO
UNC5C
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit