Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human UPB1 Partial ORF (NP_057411.1, 285 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051733Q01L

 View more versions of this product

Catalog No. 89-966-691


Add to cart

Description

Description

This gene encodes a protein that belongs to the CN hydrolase family. Beta-ureidopropionase catalyzes the last step in the pyrimidine degradation pathway. The pyrimidine bases uracil and thymine are degraded via the consecutive action of dihydropyrimidine dehydrogenase (DHPDH), dihydropyrimidinase (DHP) and beta-ureidopropionase (UP) to beta-alanine and beta-aminoisobutyric acid, respectively. UP deficiencies are associated with N-carbamyl-beta-amino aciduria and may lead to abnormalities in neurological activity. [provided by RefSeq]

Sequence: AINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYSPTIVKE
Specifications

Specifications

NP_057411.1
Liquid
51733
UPB1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BUP1
UPB1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AINRVGTEHFPNEFTSGDGKKAHQDFGYFYGSSYVAAPDSSRTPGLSRSRDGLLVAKLDLNLCQQVNDVWNFKMTGRYEMYARELAEAVKSNYSPTIVKE
RUO
UPB1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit