Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse/Rat Urotensin-2 EIA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner EIAUTS21
Optimized for serum, plasma and cell culture media. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8850