Learn More
Abnova™ Human USP16 Full-length ORF (AAH30777.1, 1 a.a. - 822 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $819.00
Specifications
Accession Number | AAH30777.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10600 |
Molecular Weight (g/mol) | 119.8kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-966-775
|
Abnova™
H00010600P01S |
10 ug |
Each for $499.00
|
|
|||||
89-966-776
|
Abnova™
H00010600P01L |
25 ug |
Each for $819.00
|
|
|||||
Description
This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MGKKRTKGKTVPIDDSSETLEPVCRHIRKGLEQGNLKKALVNVEWNICQDCKTDNKVKDKAEEETEEKPSVWLCLKCGHQGCGRNSQEQHALKHYLTPRSEPHCLVLSLDNWSVWCYVCDNEVQYCSSNQLGQVVDYVRKHASITTPKPEKDNGNIELENKKLEKESKNEQEREKKENMAKENPPMNSPCQITVKGLSNLGNTCFFNAVMQNLSQTPVLRELLKEVKMSGTIVKIEPPDLALTEPLEINLEPPGPLTLAMSQFLNEMQETKKGVVTPKELFSQVCKKAVRFKGYQQQDSQELLRYLLDGMRAEEHQRVSKGILKAFGNSTEKLDEELKNKVKDYEKKKSMPSFVDRIFGGELTSMIMCDQCRTVSLVHESFLDLSLPVLDDQSGKKSVNDKNLKKTVEDEDQDSEEEKDNDSYIKERSDIPSGTSKHLQKKAKKQAKKQAKNQRRQQKIQGKVLHLNDICTIDHPEDSDNEAEMSLQGEVNIKSNHISQEGVMHKEYCVNQKDLNGQAKMIESVTDNQKSTEEVDMKNINMDNDLEVLTSSPTRNLNGAYLTEGSNGEVDISNGFKNLNLNAALHPDEINIEILNDSHTPGTKVYEVVNEDPETAFCTLANREVFNTDECSIQHCLYQFTRNEKLRDANKLLCEVCTRRQCNGPKANIKGERKHVYTNAKKQMLISLAPPVLTLHLKRFQQAGFNLRKVNKHIKFPEILDLAPFCTLKCKNVAEENTRVLYSLYGVVEHSGTMRSGHYTAYAKARTANSHLSNLVLHGDIPQDFEMESKGQWFHISDTHVQAVPTTKVLNSQAYLLFYERILSpecifications
AAH30777.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
119.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGKKRTKGKTVPIDDSSETLEPVCRHIRKGLEQGNLKKALVNVEWNICQDCKTDNKVKDKAEEETEEKPSVWLCLKCGHQGCGRNSQEQHALKHYLTPRSEPHCLVLSLDNWSVWCYVCDNEVQYCSSNQLGQVVDYVRKHASITTPKPEKDNGNIELENKKLEKESKNEQEREKKENMAKENPPMNSPCQITVKGLSNLGNTCFFNAVMQNLSQTPVLRELLKEVKMSGTIVKIEPPDLALTEPLEINLEPPGPLTLAMSQFLNEMQETKKGVVTPKELFSQVCKKAVRFKGYQQQDSQELLRYLLDGMRAEEHQRVSKGILKAFGNSTEKLDEELKNKVKDYEKKKSMPSFVDRIFGGELTSMIMCDQCRTVSLVHESFLDLSLPVLDDQSGKKSVNDKNLKKTVEDEDQDSEEEKDNDSYIKERSDIPSGTSKHLQKKAKKQAKKQAKNQRRQQKIQGKVLHLNDICTIDHPEDSDNEAEMSLQGEVNIKSNHISQEGVMHKEYCVNQKDLNGQAKMIESVTDNQKSTEEVDMKNINMDNDLEVLTSSPTRNLNGAYLTEGSNGEVDISNGFKNLNLNAALHPDEINIEILNDSHTPGTKVYEVVNEDPETAFCTLANREVFNTDECSIQHCLYQFTRNEKLRDANKLLCEVCTRRQCNGPKANIKGERKHVYTNAKKQMLISLAPPVLTLHLKRFQQAGFNLRKVNKHIKFPEILDLAPFCTLKCKNVAEENTRVLYSLYGVVEHSGTMRSGHYTAYAKARTANSHLSNLVLHGDIPQDFEMESKGQWFHISDTHVQAVPTTKVLNSQAYLLFYERIL | |
RUO | |
USP16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10600 | |
USP16 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UBP-M | |
USP16 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.