Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human USP16 Full-length ORF (AAH30777.1, 1 a.a. - 822 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010600P01S

 View more versions of this product

Catalog No. 89-966-775


Add to cart

Description

Description

This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MGKKRTKGKTVPIDDSSETLEPVCRHIRKGLEQGNLKKALVNVEWNICQDCKTDNKVKDKAEEETEEKPSVWLCLKCGHQGCGRNSQEQHALKHYLTPRSEPHCLVLSLDNWSVWCYVCDNEVQYCSSNQLGQVVDYVRKHASITTPKPEKDNGNIELENKKLEKESKNEQEREKKENMAKENPPMNSPCQITVKGLSNLGNTCFFNAVMQNLSQTPVLRELLKEVKMSGTIVKIEPPDLALTEPLEINLEPPGPLTLAMSQFLNEMQETKKGVVTPKELFSQVCKKAVRFKGYQQQDSQELLRYLLDGMRAEEHQRVSKGILKAFGNSTEKLDEELKNKVKDYEKKKSMPSFVDRIFGGELTSMIMCDQCRTVSLVHESFLDLSLPVLDDQSGKKSVNDKNLKKTVEDEDQDSEEEKDNDSYIKERSDIPSGTSKHLQKKAKKQAKKQAKNQRRQQKIQGKVLHLNDICTIDHPEDSDNEAEMSLQGEVNIKSNHISQEGVMHKEYCVNQKDLNGQAKMIESVTDNQKSTEEVDMKNINMDNDLEVLTSSPTRNLNGAYLTEGSNGEVDISNGFKNLNLNAALHPDEINIEILNDSHTPGTKVYEVVNEDPETAFCTLANREVFNTDECSIQHCLYQFTRNEKLRDANKLLCEVCTRRQCNGPKANIKGERKHVYTNAKKQMLISLAPPVLTLHLKRFQQAGFNLRKVNKHIKFPEILDLAPFCTLKCKNVAEENTRVLYSLYGVVEHSGTMRSGHYTAYAKARTANSHLSNLVLHGDIPQDFEMESKGQWFHISDTHVQAVPTTKVLNSQAYLLFYERIL
Specifications

Specifications

AAH30777.1
Liquid
10600
USP16 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGKKRTKGKTVPIDDSSETLEPVCRHIRKGLEQGNLKKALVNVEWNICQDCKTDNKVKDKAEEETEEKPSVWLCLKCGHQGCGRNSQEQHALKHYLTPRSEPHCLVLSLDNWSVWCYVCDNEVQYCSSNQLGQVVDYVRKHASITTPKPEKDNGNIELENKKLEKESKNEQEREKKENMAKENPPMNSPCQITVKGLSNLGNTCFFNAVMQNLSQTPVLRELLKEVKMSGTIVKIEPPDLALTEPLEINLEPPGPLTLAMSQFLNEMQETKKGVVTPKELFSQVCKKAVRFKGYQQQDSQELLRYLLDGMRAEEHQRVSKGILKAFGNSTEKLDEELKNKVKDYEKKKSMPSFVDRIFGGELTSMIMCDQCRTVSLVHESFLDLSLPVLDDQSGKKSVNDKNLKKTVEDEDQDSEEEKDNDSYIKERSDIPSGTSKHLQKKAKKQAKKQAKNQRRQQKIQGKVLHLNDICTIDHPEDSDNEAEMSLQGEVNIKSNHISQEGVMHKEYCVNQKDLNGQAKMIESVTDNQKSTEEVDMKNINMDNDLEVLTSSPTRNLNGAYLTEGSNGEVDISNGFKNLNLNAALHPDEINIEILNDSHTPGTKVYEVVNEDPETAFCTLANREVFNTDECSIQHCLYQFTRNEKLRDANKLLCEVCTRRQCNGPKANIKGERKHVYTNAKKQMLISLAPPVLTLHLKRFQQAGFNLRKVNKHIKFPEILDLAPFCTLKCKNVAEENTRVLYSLYGVVEHSGTMRSGHYTAYAKARTANSHLSNLVLHGDIPQDFEMESKGQWFHISDTHVQAVPTTKVLNSQAYLLFYERIL
RUO
USP16
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
119.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UBP-M
USP16
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit