Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human USP48 Partial ORF (NP_115612, 110 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00084196Q01L

 View more versions of this product

Catalog No. 89-966-851


Add to cart

Description

Description

This gene encodes a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: NLELRQALYLCPSTCSDYMLGDGIQEEKDYEPQTICEHLQYLFALLQNSNRRYIDPSGFVKALGLDTGQQQDAQEFSKLFMSLLEDTLSKQKNPDVRNIVQQQFCGEYAY
Specifications

Specifications

NP_115612
Liquid
84196
USP48 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp762M1713|MGC132556|MGC14879|RAP1GA1|USP31
USP48
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NLELRQALYLCPSTCSDYMLGDGIQEEKDYEPQTICEHLQYLFALLQNSNRRYIDPSGFVKALGLDTGQQQDAQEFSKLFMSLLEDTLSKQKNPDVRNIVQQQFCGEYAY
RUO
USP48
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit