Learn More
Abnova™ Human VCX3A Partial ORF (NP_057463, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051481Q01S
Description
This gene belongs to the VCX/Y gene family, which has multiple members on both X and Y chromosomes, and all are expressed exclusively in male germ cells. The X-linked members are clustered on chromosome Xp22 and Y-linked members are two identical copies of the gene within a palindromic region on Yq11. The family members share a high degree of sequence identity, with the exception that a 30-bp unit is tandemly repeated in X-linked members but occurs only once in Y-linked members. The VCX gene cluster is polymorphic in terms of copy number; different individuals may have a different number of VCX genes. VCX/Y genes encode small and highly charged proteins of unknown function. The presence of a putative bipartite nuclear localization signal suggests that VCX/Y members are nuclear proteins. This gene contains 8 repeats of the 30-bp unit. [provided by RefSeq]
Sequence: SPKPRASGPPAKATEAGKRKSSSQPSPSDPKKKTTKVAKKGKAVRRGRRGKKGAATKMAAVTAPEAESGPAAPGPSDQPSQELPQHELPPEEPVSEGTQSpecifications
NP_057463 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SPKPRASGPPAKATEAGKRKSSSQPSPSDPKKKTTKVAKKGKAVRRGRRGKKGAATKMAAVTAPEAESGPAAPGPSDQPSQELPQHELPPEEPVSEGTQ | |
RUO | |
VCX3A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51481 | |
VCX3A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC118976/MGC125730/MGC125796/VCX-8r/VCX-A/VCX3/VCX8R/VCXA | |
VCX3A | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.