Learn More
R&D Systems™ Human VEGF 165 Biotinylated, Carrier-Free Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ BT293010/CF
Catalog No. BT293010CF
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human VEGF 165 Biotinylated Protein is derived from Sf 21 (baculovirus). The Recombinant Human VEGF 165 Biotinylated Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Spodoptera frugiperda, Sf 21 (baculovirus)-derived Ala27-Arg191
Bioactivity
Theoretical M.W.: 19kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
VEGFA
Biotin
Yes
NP_001165097
7422
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
<0.10 EU per 1μg of the protein by the LAL method.
VEGF
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is typically 1-6ng/mL.
Human
Reconstitute at 100μg/mL in 4mM HCl.