Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human VEGFR1/Flt-1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP191265
Description
The Human VEGFR1/Flt-1 ELISA Kit (Colorimetric) measures human VEGFR1/Flt-1 in cell culture supernatant, serum, plasma (citrate, edta).Specifications
0.16 - 10.0ng/mL | |
ELISA Kit | |
Human | |
2321 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human sVEGF-R1, Biotin-Conjugate anti-human sVEGF-R1 polyclonal antibody, Streptavidin-HRP, Human sVEGF-R1 Standard lyophilized, 20ng/mL upon reconstitution, Conjugate Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases | |
EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endot |
0.03ng/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum | |
ELISA | |
FLT1 | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
VEGFR1/Flt-1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction