Learn More
Novus Biologicals Human VG5Q ELISA Kit (Chemiluminescence)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP266659
Description
The Human VG5Q ELISA Kit (Chemiluminescence) measures human VG5Q in serum, plasma and other biological fluids.Specifications
ELISA Kit (Chemiluminescence) | |
This kit recognizes Human AGGF1/VG5Q in samples. No significant cross-reactivity or interference between Human AGGF1/VG5Q and analogues was observed. | |
55109 | |
VG5Q | |
1 Kit | |
18.75 pg/mL |
31.25-2000 pg/mL | |
ELISA | |
Storage is content dependent. | |
Human | |
Biological Fluids, Plasma, Serum | |
AGGF1 |