Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VIGR/GPR126 MAb (Clone 1044430), R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $420.50
Specifications
Antigen | VIGR/GPR126 |
---|---|
Clone | 1044430 |
Dilution | Flow Cytometry 0.25 ug/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10577SP
![]() |
R&D Systems
MAB10577SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10577100
![]() |
R&D Systems
MAB10577100 |
100 μg |
Each for $420.50
|
|
|||||
Description
VIGR/GPR126 Monoclonal antibody specifically detects VIGR/GPR126 in Human samples. It is validated for Flow CytometrySpecifications
VIGR/GPR126 | |
Flow Cytometry 0.25 ug/106 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
APG1, Developmentally regulated G-protein-coupled receptor, DREG, FLJ14937, G protein-coupled receptor 126, Gm222, GPR126, G-protein coupled receptor 126, HBV PreS1-transactivated protein 2, PS1TP2, Vascular inducible G protein-coupled receptor, vascular-inducible G protein-coupled receptor, VIGRPS1TP2 | |
Human embryonic kidney cell HEK293-derived human VIGR/GPR126 protein, Cys38-Lys437, Accession # AAH75798.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1044430 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
AAH75798.1 | |
57211 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title