Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VMAT2 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8327U100UG
This item is not returnable.
View return policy
Description
VMAT2 Monoclonal specifically detects VMAT2 in Human samples. It is validated for Flow Cytometry.Specifications
VMAT2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q05940 | |
SLC18A2 | |
NS0 mouse myeloma cell line transfected with human VMAT2 Met1-Asp514 Accession # Q05940 | |
RUO | |
6571 | |
Human | |
IgG1 |
Flow Cytometry | |
899327 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
MGC120477, MNAT, monoamine neurotransmitter transporter, Monoamine transporter, SLC18A2, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 | |
Mouse | |
100 μg | |
Primary | |
Detects human VMAT2 in ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction