Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VNN1 Full-length ORF (NP_004657.1, 1 a.a. - 513 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008876P01L

 View more versions of this product

Catalog No. 89-967-075


Add to cart

Description

Description

This gene encodes a member of the vanin family of proteins, which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. This protein, like its mouse homolog, is likely a GPI-anchored cell surface molecule. The mouse protein is expressed by the perivascular thymic stromal cells and regulates migration of T-cell progenitors to the thymus. This gene lies in close proximity to, and in the same transcriptional orientation as, two other vanin genes on chromosome 6q23-q24. [provided by RefSeq]

Sequence: MTTQLPAYVAILLFYVSRASCQDTFIAAVYEHAAILPNATLTPVSREEALALMNRNLDILEGAITSAADQGAHIIVTPEDAIYGWNFNRDSLYPYLEDIPDPEVNWIPCNNRNRFGQTPVQERLSCLAKNNSIYVVANIGDKKPCDTSDPQCPPDGRYQYNTDVVFDSQGKLVARYHKQNLFMGENQFNVPKEPEIVTFNTTFGSFGIFTCFDILFHDPAVTLVKDFHVDTIVFPTAWMNVLPHLSAVEFHSAWAMGMRVNFLASNIHYPSKKMTGSGIYAPNSSRAFHYDMKTEEGKLLLSQLDSHPSHSAVVNWTSYASSIEALSSGNKEFKGTVFFDEFTFVKLTGVAGNYTVCQKDLCCHLSYKMSENIPNEVYALGAFDGLHTVEGRYYLQICTLLKCKTTNLNTCGDSAETASTRFEMFSLSGTFGTQYVFPEVLLSENQLAPGEFQVSTDGRLFSLKPTSGPVLTVTLFGRLYEKDWASNASSGLTAQARIIMLIVIAPIVCSLSW
Specifications

Specifications

NP_004657.1
Liquid
8876
VNN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTTQLPAYVAILLFYVSRASCQDTFIAAVYEHAAILPNATLTPVSREEALALMNRNLDILEGAITSAADQGAHIIVTPEDAIYGWNFNRDSLYPYLEDIPDPEVNWIPCNNRNRFGQTPVQERLSCLAKNNSIYVVANIGDKKPCDTSDPQCPPDGRYQYNTDVVFDSQGKLVARYHKQNLFMGENQFNVPKEPEIVTFNTTFGSFGIFTCFDILFHDPAVTLVKDFHVDTIVFPTAWMNVLPHLSAVEFHSAWAMGMRVNFLASNIHYPSKKMTGSGIYAPNSSRAFHYDMKTEEGKLLLSQLDSHPSHSAVVNWTSYASSIEALSSGNKEFKGTVFFDEFTFVKLTGVAGNYTVCQKDLCCHLSYKMSENIPNEVYALGAFDGLHTVEGRYYLQICTLLKCKTTNLNTCGDSAETASTRFEMFSLSGTFGTQYVFPEVLLSENQLAPGEFQVSTDGRLFSLKPTSGPVLTVTLFGRLYEKDWASNASSGLTAQARIIMLIVIAPIVCSLSW
RUO
VNN1
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HDLCQ8|MGC116930|MGC116931|MGC116932|MGC116933|Tiff66
VNN1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit