Learn More
Abnova™ Human VNN2 Full-length ORF (NP_511043.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00008875P01L
Description
This gene product is a member of the Vanin family of proteins which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MNENIDILETAIKQAAEQGARIIVTPEDALYGWKFTRETVFPYLEDIPDPQVNWIPCQDPHRFGHTPVQARLSCLAKDNSIYVLANLGDKKPCNSRDSTCPPNGYFQYNTNVVYNTEGKLVARYHKYHLYSEPQFNVPEKPELVTFNTAFGRFGIFTCFDIFFYDPGVTLVKDFHVDTILFPTAWMNVLPLLTAIEFHSAWAMGMGVNLLVANTHHVSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSCGTSNSAITYLLIFILLMIIALQNIVMLSpecifications
NP_511043.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FOAP-4/GPI-80 | |
VNN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8875 | |
VNN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNENIDILETAIKQAAEQGARIIVTPEDALYGWKFTRETVFPYLEDIPDPQVNWIPCQDPHRFGHTPVQARLSCLAKDNSIYVLANLGDKKPCNSRDSTCPPNGYFQYNTNVVYNTEGKLVARYHKYHLYSEPQFNVPEKPELVTFNTAFGRFGIFTCFDIFFYDPGVTLVKDFHVDTILFPTAWMNVLPLLTAIEFHSAWAMGMGVNLLVANTHHVSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSCGTSNSAITYLLIFILLMIIALQNIVML | |
RUO | |
VNN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.