Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VNN2 Partial ORF (NP_004656, 320 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008875Q01L

 View more versions of this product

Catalog No. 89-967-079


Add to cart

Description

Description

This gene product is a member of the Vanin family of proteins which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: WNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPV
Specifications

Specifications

NP_004656
Liquid
8875
VNN2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FOAP-4|GPI-80
VNN2
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPV
RUO
VNN2
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit