Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPREB3 Full-length ORF (NP_037510.1, 1 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name VPREB3 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 40.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-967-090
View Documents
Abnova™
H00029802P01L
25 ug
Each for $709.50
Add to cart
 
89-967-089
View Documents
Abnova™
H00029802P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The protein encoded by this gene is the human homolog of the mouse VpreB3 (8HS20) protein, and is specifically expressed in cell lines representative of all stages of B-cell differentiation. It is also related to VPREB1 and other members of the immunoglobulin supergene family. This protein associates with membrane mu heavy chains early in the course of pre-B cell receptor biosynthesis. The precise function of the protein is not known, but it may contribute to mu chain transport in pre-B cells. [provided by RefSeq]

Sequence: MACRCLSFLLMGTFLSVSQTVLAQLDALLVFPGQVAQLSCTLSPQHVTIRDYGVSWYQQRAGSAPRYLLYYRSEEDHHRPADIPDRFSAAKDEAHNACVLTISPVQPEDDADYYCSVGYGFSP
Specifications

Specifications

VPREB3 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
40.1kDa
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit