Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS16 Full-length ORF (NP_072097.2, 1 a.a. - 839 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064601P01L

 View more versions of this product

Catalog No. 89-967-099


Add to cart

Description

Description

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps16 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MDCYTANWNPLGDSAFYRKYELYSMDWDLKEELRDCLVAAAPYGGPIALLRNPWRKEKAASVRPVLDIYSASGMPLASLLWKSGPVVSLGWSAEEELLCVQEDGAVLVYGLHGDFRRHFSMGNEVLQNRVLDARIFHTEFGSGVAILTGAHRFTLSANVGDLKLRRMPEVPGLQSAPSCWTVLCQDRVAHILLAVGPDLYLLDHAACSAVTPPGLAPGVSSFLQMAVSFTYRHLALFTDTGYIWMGTASLKEKLCEFNCNIRAPPKQMVWCSRPRSKERAVVVAWERRLMVVGDAPESIQFVLDEDSYLVPELDGVRIFSRSTHEFLHEVPAASEEIFKIASMAPGALLLEAQKEYEKESQKADEYLREIQELGQLTQAVQQCIEAAGHEHQPDMQKSLLRAASFGKCFLDRFPPDSFVHMCQDLRVLNAVRDYHIGIPLTYSQYKQLTIQVLLDRLVLRRLYPLAIQICEYLRLPEVQGVSRILAHWACYKVQQKDVSDEDVARAINQKLGDTPGVSYSDIAARAYGCGRTELAIKLLEYEPRSGEQVPLLLKMKRSKLALSKAIESGDTDLVFTVLLHLKNELNRGDFFMTLRNQPMALSLYRQFCKHQELETLKDLYNQDDNHQELGSFHIRASYAAEERIEGRVAALQTAADAFYKAKNEFAAKATEDQMRLLRLQRRLEDELGGQFLDLSLHDTVTTLILGGHNKRAEQLARDFRIPDKRLWWLKLTALADLEDWEELEKFSKSKKSPIGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKK
Specifications

Specifications

NP_072097.2
Liquid
64601
VPS16 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDCYTANWNPLGDSAFYRKYELYSMDWDLKEELRDCLVAAAPYGGPIALLRNPWRKEKAASVRPVLDIYSASGMPLASLLWKSGPVVSLGWSAEEELLCVQEDGAVLVYGLHGDFRRHFSMGNEVLQNRVLDARIFHTEFGSGVAILTGAHRFTLSANVGDLKLRRMPEVPGLQSAPSCWTVLCQDRVAHILLAVGPDLYLLDHAACSAVTPPGLAPGVSSFLQMAVSFTYRHLALFTDTGYIWMGTASLKEKLCEFNCNIRAPPKQMVWCSRPRSKERAVVVAWERRLMVVGDAPESIQFVLDEDSYLVPELDGVRIFSRSTHEFLHEVPAASEEIFKIASMAPGALLLEAQKEYEKESQKADEYLREIQELGQLTQAVQQCIEAAGHEHQPDMQKSLLRAASFGKCFLDRFPPDSFVHMCQDLRVLNAVRDYHIGIPLTYSQYKQLTIQVLLDRLVLRRLYPLAIQICEYLRLPEVQGVSRILAHWACYKVQQKDVSDEDVARAINQKLGDTPGVSYSDIAARAYGCGRTELAIKLLEYEPRSGEQVPLLLKMKRSKLALSKAIESGDTDLVFTVLLHLKNELNRGDFFMTLRNQPMALSLYRQFCKHQELETLKDLYNQDDNHQELGSFHIRASYAAEERIEGRVAALQTAADAFYKAKNEFAAKATEDQMRLLRLQRRLEDELGGQFLDLSLHDTVTTLILGGHNKRAEQLARDFRIPDKRLWWLKLTALADLEDWEELEKFSKSKKSPIGYLPFVEICMKQHNKYEAKKYASRVGPEQKVKALLLVGDVAQAADVAIEHRNEAELSLVLSHCTGATDGATADKIQRARAQAQKK
RUO
VPS16
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
121.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
hVPS16
VPS16
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit