Learn More
Abnova™ Human VPS18 Full-length ORF (AAH01513.1, 1 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00057617P01L
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps18 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. [provided by RefSeq]
Sequence: MASILDEYENSLSRSAVLQPGCPSVGIPHSGYVNAQLEKEVPIFTKQRIDFTPSERITSLVVSSNQLCMSLGKDTLLRIDLGKANEPNHVELGRKDDAKVHKMFLDHTGSHLLIALSSTEVLYVNRNGQKVRPLARWKGQLVESVGWNKALGTESSTGPILVGTAQGHIFEAELSASEGGLFGPAPDLYFRPLYVLNEEGGPAPVCSLEAERGPDGRSFVIATTRQRLFQFIGRAAEGAEAQGFSGLFAAYTDHPPPFREFPSNLGYSELAFYTPKLRSAPRAFAWMMGDGVLYGALDCGRPDSLLSEERVWEYPEGVGPGASPPLAIVLTQFHFLLLLADRVEAVCTLTGQVVLRDHFLEKFGPLKHMVKDSSTGQLWAYTERAVFRYHVQREARDVWRTYLDMNRFDLAKEYCRERPDCLDTVLAREADFCFRQRRYLESARCYALTQSYFEEIALKFLEARQEEALAEFLQRKLASLKPAERTQATLLTTWLTELYLSRLGALQGDPEALTLYREVRNLTQFHPLPLAPLLSLSFPTHVLFTSREREREHLSSVCSLCGLWNPSSSLSEEAFSSSCLSpecifications
AAH01513.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1475/PEP3 | |
VPS18 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57617 | |
VPS18 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASILDEYENSLSRSAVLQPGCPSVGIPHSGYVNAQLEKEVPIFTKQRIDFTPSERITSLVVSSNQLCMSLGKDTLLRIDLGKANEPNHVELGRKDDAKVHKMFLDHTGSHLLIALSSTEVLYVNRNGQKVRPLARWKGQLVESVGWNKALGTESSTGPILVGTAQGHIFEAELSASEGGLFGPAPDLYFRPLYVLNEEGGPAPVCSLEAERGPDGRSFVIATTRQRLFQFIGRAAEGAEAQGFSGLFAAYTDHPPPFREFPSNLGYSELAFYTPKLRSAPRAFAWMMGDGVLYGALDCGRPDSLLSEERVWEYPEGVGPGASPPLAIVLTQFHFLLLLADRVEAVCTLTGQVVLRDHFLEKFGPLKHMVKDSSTGQLWAYTERAVFRYHVQREARDVWRTYLDMNRFDLAKEYCRERPDCLDTVLAREADFCFRQRRYLESARCYALTQSYFEEIALKFLEARQEEALAEFLQRKLASLKPAERTQATLLTTWLTELYLSRLGALQGDPEALTLYREVRNLTQFHPLPLAPLLSLSFPTHVLFTSREREREHLSSVCSLCGLWNPSSSLSEEAFSSSCL | |
RUO | |
VPS18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.