Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS33B Full-length ORF (AAH16445.1, 1 a.a. - 617 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026276P01S

 View more versions of this product

Catalog No. 89-967-124


Add to cart

Description

Description

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. [provided by RefSeq]

Sequence: MAFPHRPDAPELPDFSMLKRLARDQLIYLLEQLPGKKDLFIEADLMSPLDRIANVSILKQHEVDKLYKVENKPALSSNEQLCFLVRPRIKNMRYIASLVNADKLAGRTRKYKVIFSPQKFYACEMVLEEEGIYGDVSCDEWAFSLLPLDVDLLSMELPEFFRDYFLEGDQRWINTVAQALHLLSTLYGPFPNCYGIGRCAKMAYELWRNLEEEEDGETKGRRPEIGHIFLLDRDVDFVTALCSQVVYEGLVDDTFRIKCGSVDFGPEVTSSDKSLKVLLNAEDKVFNEIRNEHFSNVFGFLSQKARNLQAQYDRRRGMDIKQMKNFVSQELKGLKQEHRLLSLHIGACESIMKKKTKQDFQELIKTEHALLEGFNIRESTSYIEEHIDRQVSPIESLRLMCLLSITENGLIPKDYRSLKTQYLQSYGPEHLLTFSNLRRAGLLTEQAPGDTLTAVESKVSKLVTDKAAGKITDAFSSLAKRSNFRAISKKLNLIPRVDGEYDLKVPRDMAYVFSGAYVPLSCRIIEQVLERRSWQGLDEVVRLLNCSDFAFTDMTKEDKASSESLRLILVVFLGGCTFSEISALRFLGREKGYRFIFLTTAVTNSARLMEAMSEVKA
Specifications

Specifications

AAH16445.1
Liquid
26276
VPS33B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAFPHRPDAPELPDFSMLKRLARDQLIYLLEQLPGKKDLFIEADLMSPLDRIANVSILKQHEVDKLYKVENKPALSSNEQLCFLVRPRIKNMRYIASLVNADKLAGRTRKYKVIFSPQKFYACEMVLEEEGIYGDVSCDEWAFSLLPLDVDLLSMELPEFFRDYFLEGDQRWINTVAQALHLLSTLYGPFPNCYGIGRCAKMAYELWRNLEEEEDGETKGRRPEIGHIFLLDRDVDFVTALCSQVVYEGLVDDTFRIKCGSVDFGPEVTSSDKSLKVLLNAEDKVFNEIRNEHFSNVFGFLSQKARNLQAQYDRRRGMDIKQMKNFVSQELKGLKQEHRLLSLHIGACESIMKKKTKQDFQELIKTEHALLEGFNIRESTSYIEEHIDRQVSPIESLRLMCLLSITENGLIPKDYRSLKTQYLQSYGPEHLLTFSNLRRAGLLTEQAPGDTLTAVESKVSKLVTDKAAGKITDAFSSLAKRSNFRAISKKLNLIPRVDGEYDLKVPRDMAYVFSGAYVPLSCRIIEQVLERRSWQGLDEVVRLLNCSDFAFTDMTKEDKASSESLRLILVVFLGGCTFSEISALRFLGREKGYRFIFLTTAVTNSARLMEAMSEVKA
RUO
VPS33B
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14848
VPS33B
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit