Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS35 Full-length ORF (NP_060676.2, 1 a.a. - 796 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00055737P01L

 View more versions of this product

Catalog No. 89-967-130


Add to cart

Description

Description

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. [provided by RefSeq]

Sequence: MPTTQQSPQDEQEKLLDEAIQAVKVQSFQMKRCLDKNKLMDALKHASNMLGELRTSMLSPKSYYELYMAISDELHYLEVYLTDEFAKGRKVADLYELVQYAGNIIPRLYLLITVGVVYVKSFPQSRKDILKDLVEMCRGVQHPLRGLFLRNYLLQCTRNILPDEGEPTDEETTGDISDSMDFVLLNFAEMNKLWVRMQHQGHSRDREKRERERQELRILVGTNLVRLSQLEGVNVERYKQIVLTGILEQVVNCRDALAQEYLMECIIQVFPDEFHLQTLNPFLRACAELHQNVNVKNIIIALIDRLALFAHREDGPGIPADIKLFDIFSQQVATVIQSRQDMPSEDVVSLQVSLINLAMKCYPDRVDYVDKVLETTVEIFNKLNLEHIATSSAVSKELTRLLKIPVDTYNNILTVLKLKHFHPLFEYFDYESRKSMSCYVLSNVLDYNTEIVSQDQVDSIMNLVSTLIQDQPDQPVEDPDPEDFADEQSLVGRFIHLLRSEDPDQQYLILNTARKHFGAGGNQRIRFTLPPLVFAAYQLAFRYKENSKVDDKWEKKCQKIFSFAHQTISALIKAELAELPLRLFLQGALAAGEIGFENHETVAYEFMSQAFSLYEDEISDSKAQLAAITLIIGTFERMKCFSEENHEPLRTQCALAASKLLKKPDQGRAVSTCAHLFWSGRNTDKNGEELHGGKRVMECLKKALKIANQCMDPSLQVQLFIEILNRYIYFYEKENDAVTIQVLNQLIQKIREDLPNLESSEETEQINKHFHNTLEHLRLRRESPESEGPIYEGLIL
Specifications

Specifications

NP_060676.2
Liquid
55737
VPS35 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPTTQQSPQDEQEKLLDEAIQAVKVQSFQMKRCLDKNKLMDALKHASNMLGELRTSMLSPKSYYELYMAISDELHYLEVYLTDEFAKGRKVADLYELVQYAGNIIPRLYLLITVGVVYVKSFPQSRKDILKDLVEMCRGVQHPLRGLFLRNYLLQCTRNILPDEGEPTDEETTGDISDSMDFVLLNFAEMNKLWVRMQHQGHSRDREKRERERQELRILVGTNLVRLSQLEGVNVERYKQIVLTGILEQVVNCRDALAQEYLMECIIQVFPDEFHLQTLNPFLRACAELHQNVNVKNIIIALIDRLALFAHREDGPGIPADIKLFDIFSQQVATVIQSRQDMPSEDVVSLQVSLINLAMKCYPDRVDYVDKVLETTVEIFNKLNLEHIATSSAVSKELTRLLKIPVDTYNNILTVLKLKHFHPLFEYFDYESRKSMSCYVLSNVLDYNTEIVSQDQVDSIMNLVSTLIQDQPDQPVEDPDPEDFADEQSLVGRFIHLLRSEDPDQQYLILNTARKHFGAGGNQRIRFTLPPLVFAAYQLAFRYKENSKVDDKWEKKCQKIFSFAHQTISALIKAELAELPLRLFLQGALAAGEIGFENHETVAYEFMSQAFSLYEDEISDSKAQLAAITLIIGTFERMKCFSEENHEPLRTQCALAASKLLKKPDQGRAVSTCAHLFWSGRNTDKNGEELHGGKRVMECLKKALKIANQCMDPSLQVQLFIEILNRYIYFYEKENDAVTIQVLNQLIQKIREDLPNLESSEETEQINKHFHNTLEHLRLRRESPESEGPIYEGLIL
RUO
VPS35
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434E1211|DKFZp434P1672|FLJ10752|FLJ13588|FLJ20388|MEM3
VPS35
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit