Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS52 Full-length ORF (NP_072047.4, 1 a.a. - 723 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00006293P01L

 View more versions of this product

Catalog No. 89-967-153


Add to cart

Description

Description

This gene encodes a protein that is similar to the yeast suppressor of actin mutations 2 gene. The yeast protein forms a subunit of the tetrameric Golgi-associated retrograde protein complex that is involved in vesicle trafficking from from both early and late endosomes, back to the trans-Golgi network. This gene is located on chromosome 6 in a head-to-head orientation with the gene encoding ribosomal protein S18. [provided by RefSeq]

Sequence: MAAAATMAAAARELVLRAGTSDMEEEEGPLAGGPGLQEPLQLGELDITSDEFILDEVDVHIQANLEDELVKEALKTGVDLRHYSKQVELELQQIEQKSIRDYIQESENIASLHNQITACDAVLERMEQMLGAFQSDLSSISSEIRTLQEQSGAMNIRLRNRQAVRGKLGELVDGLVVPSALVTAILEAPVTEPRFLEQLQELDAKAAAVREQEARGTAACADVRGVLDRLRVKAVTKIREFILQKIYSFRKPMTNYQIPQTALLKYRFFYQFLLGNERATAKEIRDEYVETLSKIYLSYYRSYLGRLMKVQYEEVAEKDDLMGVEDTAKKGFFSKPSLRSRNTIFTLGTRGSVISPTELEAPILVPHTAQRGEQRYPFEALFRSQHYALLDNSCREYLFICEFFVVSGPAAHDLFHAVMGRTLSMTLKHLDSYLADCYDAIAVFLCIHIVLRFRNIAAKRDVPALDRYWEQVLALLWPRFELILEMNVQSVRSTDPQRLGGLDTRPHYITRRYAEFSSALVSINQTIPNERTMQLLGQLQVEVENFVLRVAAEFSSRKEQLVFLINNYDMMLGVLMERAADDSKEVESFQQLLNARTQEFIEELLSPPFGGLVAFVKEAEALIERGQAERLRGEEARVTQLIRGFGSSWKSSVESLSQDVMRSFTNFRNGTSIIQGALTQLIQLYHRFHRVLSQPQLRALPARAELINIHHLMVELKKHKPNF
Specifications

Specifications

NP_072047.4
Liquid
6293
VPS52 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAATMAAAARELVLRAGTSDMEEEEGPLAGGPGLQEPLQLGELDITSDEFILDEVDVHIQANLEDELVKEALKTGVDLRHYSKQVELELQQIEQKSIRDYIQESENIASLHNQITACDAVLERMEQMLGAFQSDLSSISSEIRTLQEQSGAMNIRLRNRQAVRGKLGELVDGLVVPSALVTAILEAPVTEPRFLEQLQELDAKAAAVREQEARGTAACADVRGVLDRLRVKAVTKIREFILQKIYSFRKPMTNYQIPQTALLKYRFFYQFLLGNERATAKEIRDEYVETLSKIYLSYYRSYLGRLMKVQYEEVAEKDDLMGVEDTAKKGFFSKPSLRSRNTIFTLGTRGSVISPTELEAPILVPHTAQRGEQRYPFEALFRSQHYALLDNSCREYLFICEFFVVSGPAAHDLFHAVMGRTLSMTLKHLDSYLADCYDAIAVFLCIHIVLRFRNIAAKRDVPALDRYWEQVLALLWPRFELILEMNVQSVRSTDPQRLGGLDTRPHYITRRYAEFSSALVSINQTIPNERTMQLLGQLQVEVENFVLRVAAEFSSRKEQLVFLINNYDMMLGVLMERAADDSKEVESFQQLLNARTQEFIEELLSPPFGGLVAFVKEAEALIERGQAERLRGEEARVTQLIRGFGSSWKSSVESLSQDVMRSFTNFRNGTSIIQGALTQLIQLYHRFHRVLSQPQLRALPARAELINIHHLMVELKKHKPNF
RUO
VPS52
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARE1|DKFZp547I194|RP5-1033B10|SAC2|SACM2L|dJ1033B10.5
VPS52
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit