Learn More
Abnova™ Human VRK3 Full-length ORF (AAH23556.1, 1 a.a. - 412 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051231P01S
Description
This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq]
Sequence: MISFCPDCGKSIQAAFKFCPYCGNSLPVEEHVGSQTFVNPHVSSFQGSKRGLNSSFETSPKKVKWSSTVTSPRLSLFSDGDSSESEDTLSSSERSKGSGSRPPTPKSSPQKTRKSPQVTRGSPQKTSCSPQKTRQSPQTLKRSRVTTSLEALPTGTVLTDKSGRQWKLKSFQTRDNQGILYEAAPTSTLTCDSGPQKQKFSLKLDAKDGRLFNEQNFFQRAAKPLQVNKWKKLYSTPLLAIPTCMGFGVHQDKYRFLVLPSLGRSLQSALDVSPKHVLSERSVLQVACRLLDALEFLHENEYVHGNVTAENIFVDPEDQSQVTLAGYGFAFRYCPSGKHVAYVEGSRSPHEGDLEFISMDLHKGCGPSRRSDLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKLPWDSFSpecifications
AAH23556.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VRK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51231 | |
VRK3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MISFCPDCGKSIQAAFKFCPYCGNSLPVEEHVGSQTFVNPHVSSFQGSKRGLNSSFETSPKKVKWSSTVTSPRLSLFSDGDSSESEDTLSSSERSKGSGSRPPTPKSSPQKTRKSPQVTRGSPQKTSCSPQKTRQSPQTLKRSRVTTSLEALPTGTVLTDKSGRQWKLKSFQTRDNQGILYEAAPTSTLTCDSGPQKQKFSLKLDAKDGRLFNEQNFFQRAAKPLQVNKWKKLYSTPLLAIPTCMGFGVHQDKYRFLVLPSLGRSLQSALDVSPKHVLSERSVLQVACRLLDALEFLHENEYVHGNVTAENIFVDPEDQSQVTLAGYGFAFRYCPSGKHVAYVEGSRSPHEGDLEFISMDLHKGCGPSRRSDLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKLPWDSF | |
RUO | |
VRK3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.