Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG8 Alexa Fluor 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9418U100UG
This item is not returnable.
View return policy
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry.Specifications
VSIG8 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
V-set and immunoglobulin domain containing 8 | |
Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13 | |
100 μg | |
391123 | |
Human | |
Supernatant |
Flow Cytometry | |
961823 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q5VU13 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction