Learn More
Abnova™ Human Vsig8 Full-Length Orf (Adr83364.1) Recombinant Protein Without Tag.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Supplier: Abnova™ H00391123G01
Description
Good FoldingImmunogen Sequence: MRVGGAFHLLLVCLSPALLSAVRINGDGQEVLYLAEGDNVRLGCPYVLDPEDYGPNGLDIEWMQVNSDPAHHRENVFLSYQDKRINHGSLPHLQQRVRFAASDPSQYDASINLMNLQVSDTATYECRVKKTTMATRKVIVTVQARPAVPMCWTEGHMTYGNDVVLKCYASGGSQPLSYKWAKISGHHYPYRAGSYTSQHSYHS
Specifications
ADR83364.1 | |
Liquid | |
VSIG8 (Human) Recombinant Protein | |
MRVGGAFHLLLVCLSPALLSAVRINGDGQEVLYLAEGDNVRLGCPYVLDPEDYGPNGLDIEWMQVNSDPAHHRENVFLSYQDKRINHGSLPHLQQRVRFAASDPSQYDASINLMNLQVSDTATYECRVKKTTMATRKVIVTVQARPAVPMCWTEGHMTYGNDVVLKCYASGGSQPLSYKWAKISGHHYPYRAGSYTSQHSYHSELSYQESFHSSINQGLNNGDLVLKDISRADDGLYQCTVANNVGYSVCVVEVKVSDSRRIGVIIGIVLGSLLALGCLAVGIWGLVCCCCGGSGAGGARGAFGYGNGGGVGGGACGDLASEIREDAVAPGCKASGRGSRVTHLLGYPTQNVSRSLRRKYAPPPCGGPEDVALAPCTAAAACEAGPSPVYVKVKSAEPADCAEGPVQCKNGLLV | |
RUO | |
VSIG8 | |
None |
Antibody Production, Compound Screening, Functional Study | |
391123 | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VSIG8 | |
Wheat Germ (in vitro) | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only