Learn More
Haematologic Technologies Inc HUMAN VWF 100UG
Supplier:  Haematologic Technologies Inc HCVWF0190
Encompass_Preferred
Coagulation protein, Human von Willebrand Factor, 100 micrograms, Purified from platelet plasma, supplied in 25mM Citrate, 100mM NaCl, 100mM Glycine, pH 6.8, store at -80 Celsius.
Catalog No. 50-883-482
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.