Learn More
Abnova™ Human WARS2 Partial ORF (NP_056651, 113 a.a. - 222 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010352Q01L
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: QQSQVSEHTQLSWILSCMVRLPRLQHLHQWKAKTTKQKHDGTVGLLTYPVLQAADILLYKSTHVPVGEDQVQHMELVQDLAQGFNKKYGEFFPVPESILTSMKKVKSLRDSpecifications
NP_056651 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QQSQVSEHTQLSWILSCMVRLPRLQHLHQWKAKTTKQKHDGTVGLLTYPVLQAADILLYKSTHVPVGEDQVQHMELVQDLAQGFNKKYGEFFPVPESILTSMKKVKSLRD | |
RUO | |
WARS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10352 | |
WARS2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TrpRS | |
WARS2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.