Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WASF2 Full-length ORF (NP_008921.1, 1 a.a. - 498 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010163P01L

 View more versions of this product

Catalog No. 89-967-216


Add to cart

Description

Description

This gene encodes a member of the Wiskott-Aldrich syndrome protein family. The gene product is a protein that forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function. The published map location (PMID:10381382) has been changed based on recent genomic sequence comparisons, which indicate that the expressed gene is located on chromosome 1, and a pseudogene may be located on chromosome X. [provided by RefSeq]

Sequence: MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD
Specifications

Specifications

NP_008921.1
Liquid
10163
WASF2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPLVTRNIEPRHLCRQTLPSVRSELECVTNITLANVIRQLGSLSKYAEDIFGELFTQANTFASRVSSLAERVDRLQVKVTQLDPKEEEVSLQGINTRKAFRSSTIQDQKLFDRNSLPVPVLETYNTCDTPPPLNNLTPYRDDGKEALKFYTDPSYFFDLWKEKMLQDTKDIMKEKRKHRKEKKDNPNRGNVNPRKIKTRKEEWEKMKMGQEFVESKEKLGTSGYPPTLVYQNGSIGCVENVDASSYPPPPQSDSASSPSPSFSEDNLPPPPAEFSYPVDNQRGSGLAGPKRSSVVSPSHPPPAPPLGSPPGPKPGFAPPPAPPPPPPPMIGIPPPPPPVGFGSPGTPPPPSPPSFPPHPDFAAPPPPPPPPAADYPTLPPPPLSQPTGGAPPPPPPPPPPGPPPPPFTGADGQPAIPPPLSDTTKPKSSLPAVSDARSDLLSAIRQGFQLRRVEEQREQEKRDVVGNDVATILSRRIAVEYSDSEDDSSEFDEDDWSD
RUO
WASF2
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCAR2|WAVE2|dJ393P12.2
WASF2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit