Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WASPIP Full-length ORF (AAH02914.1, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00007456P01S

 View more versions of this product

Catalog No. 89-967-224


Add to cart

Description

Description

This gene encodes a protein that plays an important role in the organization of the actin cytoskeleton. The encoded protein binds to a region of Wiskott-Aldrich syndrome protein that is frequently mutated in Wiskott-Aldrich syndrome, an X-linked recessive disorder. Impairment of the interaction between these two proteins may contribute to the disease. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]

Sequence: MPVPPPPAPPPPPTFALANTEKPTLNKTEQAGRNALLSDISKGKKLKKTVTNDRSAPILDKPKGAGAGGGGGGFGGGGGFGGGGGGGGGGSFGGGGPPGLGGLFQAGMPKLRSTANRDNHSGGSRPPLLPPGGRSTSAKPFSPPSGPGRFPVPSPGHRSGPPEPQRNRMPPPRPDVGSKPDSIPPPVPSTPRPIQSSLHNRGSPPVPGGPRQPSPGPTPPPPPVRDPPGRSGPLPPPPPVSRNGSTSRALPATPQLPSRSGVDSPRSGPRPPLPPDRPSAGAPPPPPPSTSIRNGFQDSPCEDEWESRFYFHPISDLPPPEPYVQTTKSYPSKLARNESRSGSNRRERGAPPLPPIPR
Specifications

Specifications

AAH02914.1
Liquid
7456
WASPIP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPVPPPPAPPPPPTFALANTEKPTLNKTEQAGRNALLSDISKGKKLKKTVTNDRSAPILDKPKGAGAGGGGGGFGGGGGFGGGGGGGGGGSFGGGGPPGLGGLFQAGMPKLRSTANRDNHSGGSRPPLLPPGGRSTSAKPFSPPSGPGRFPVPSPGHRSGPPEPQRNRMPPPRPDVGSKPDSIPPPVPSTPRPIQSSLHNRGSPPVPGGPRQPSPGPTPPPPPVRDPPGRSGPLPPPPPVSRNGSTSRALPATPQLPSRSGVDSPRSGPRPPLPPDRPSAGAPPPPPPSTSIRNGFQDSPCEDEWESRFYFHPISDLPPPEPYVQTTKSYPSKLARNESRSGSNRRERGAPPLPPIPR
RUO
WIPF1
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111041|PRPL-2|WASPIP|WIP
WIPF1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit