Learn More
Abnova™ Human WBP11 Full-length ORF (NP_057396.1, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051729P01L
Description
This gene encodes a nuclear protein, which colocalizes with mRNA splicing factors and intermediate filament-containing perinuclear networks. This protein has 95% amino acid sequence identity to the mouse Wbp11 protein. It contains two proline-rich regions that bind to the WW domain of Npw38, a nuclear protein, and thus this protein is also called Npw38-binding protein NpwBP. The Npw38-NpwBP complex may function as a component of an mRNA factory in the nucleus. [provided by RefSeq]
Sequence: MGRRSTSSTKSGKFMNPTDQARKEARKRELKKNKKQRMMVRAAVLKMKDPKQIIRDMEKLDEMEFNPVQQPQLNEKVLKDKRKKLRETFERILRLYEKENPDIYKELRKLEVEYEQKRAQLSQYFDAVKNAQHVEVESIPLPDMPHAPSNILIQDIPLPGAQPPSILKKTSAYGPPTRAVSILPLLGHGVPRLPPGRKPPGPPPGPPPPQVVQMYGRKVGFALDLPPRRRDEDMLYSPELAQRGHDDDVSSTSEDDGYPEDMDQDKHDDSTDDSDTDKSDGESDGDEFVHRDNGERDNNEEKKSGLSVRFADMPGKSRKKKKNMKELTPLQAMMLRMAGQEIPEEGREVEEFSEDDDEDDSDDSEAEKQSQKQHKEESHSDGTSTASSQQQAPPQSVPPSQIQAPPMPGPPPLGPPPAPPLRPPGPPTGLPPGPPPGAPPFLRPPGMPGLRGPLPRLLPPGPPPGRPPGPPPGPPPGLPPGPPPRGPPPRLPPPAPPGIPPPRPGMMRPPLVPPLGPAPPGLFPPAPLPNPGVLSAPPNLIQRPKADDTSAATIEKKATATISAKPQITNPKAEITRFVPTALRVRRENKGATAAPQRKSEDDSAVPLAKAAPKSGPSVPVSVQTKDDVYEAFMKEMEGLLSpecifications
NP_057396.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
96.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779M1063/NPWBP/SIPP1 | |
WBP11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51729 | |
WBP11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGRRSTSSTKSGKFMNPTDQARKEARKRELKKNKKQRMMVRAAVLKMKDPKQIIRDMEKLDEMEFNPVQQPQLNEKVLKDKRKKLRETFERILRLYEKENPDIYKELRKLEVEYEQKRAQLSQYFDAVKNAQHVEVESIPLPDMPHAPSNILIQDIPLPGAQPPSILKKTSAYGPPTRAVSILPLLGHGVPRLPPGRKPPGPPPGPPPPQVVQMYGRKVGFALDLPPRRRDEDMLYSPELAQRGHDDDVSSTSEDDGYPEDMDQDKHDDSTDDSDTDKSDGESDGDEFVHRDNGERDNNEEKKSGLSVRFADMPGKSRKKKKNMKELTPLQAMMLRMAGQEIPEEGREVEEFSEDDDEDDSDDSEAEKQSQKQHKEESHSDGTSTASSQQQAPPQSVPPSQIQAPPMPGPPPLGPPPAPPLRPPGPPTGLPPGPPPGAPPFLRPPGMPGLRGPLPRLLPPGPPPGRPPGPPPGPPPGLPPGPPPRGPPPRLPPPAPPGIPPPRPGMMRPPLVPPLGPAPPGLFPPAPLPNPGVLSAPPNLIQRPKADDTSAATIEKKATATISAKPQITNPKAEITRFVPTALRVRRENKGATAAPQRKSEDDSAVPLAKAAPKSGPSVPVSVQTKDDVYEAFMKEMEGLL | |
RUO | |
WBP11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.