Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDR1 Full-length ORF (NP_059830.1, 1 a.a. - 606 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009948P01L

 View more versions of this product

Catalog No. 89-967-267


Add to cart

Description

Description

This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPYEIKKVFASLPQVERGVSKIIGGDPKGNNFLYTNGKCVILRNIDNPALADIYTEHAHQVVVAKYAPSGFYIASGDVSGKLRIWDTTQKEHLLKYEYQPFAGKIKDIAWTEDSKRIAVVGEGREKFGAVFLWDSGSSVGEITGHNKVINSVDIKQSRPYRLATGSDDNCAAFFEGPPFKFKFTIGDHSRFVNCVRFSPDGNRFATASADGQIYIYDGKTGEKVCALGGSKAHDGGIYAISWSPDSTHLLSASGDKTSKIWDVSVNSVVSTFPMGSTVLDQQLGCLWQKDHLLSVSLSGYINYLDRNNPSKPLHVIKGHSKSIQCLTVHKNGGKSYIYSGSHDGHINYWDSETGENDSFAGKGHTNQVSRMTVDESGQLISCSMDDTVRYTSLMLRDYSGQGVVKLDVQPKCVAVGPGGYAVVVCIGQIVLLKDQRKCFSIDNPGYEPEVVAVHPGGDTVAIGGVDGNVRLYSILGTTLKDEGKLLEAKGPVTDVAYSHDGAFLAVCDASKVVTVFSVADGYSENNVFYGHHAKIVCLAWSPDNEHFASGGMDMMVYVWTLSDPETRVKIQDAHRLHHVSSLAWLDEHTLVTTSHDASVKEWTITY
Specifications

Specifications

NP_059830.1
Liquid
9948
WDR1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPYEIKKVFASLPQVERGVSKIIGGDPKGNNFLYTNGKCVILRNIDNPALADIYTEHAHQVVVAKYAPSGFYIASGDVSGKLRIWDTTQKEHLLKYEYQPFAGKIKDIAWTEDSKRIAVVGEGREKFGAVFLWDSGSSVGEITGHNKVINSVDIKQSRPYRLATGSDDNCAAFFEGPPFKFKFTIGDHSRFVNCVRFSPDGNRFATASADGQIYIYDGKTGEKVCALGGSKAHDGGIYAISWSPDSTHLLSASGDKTSKIWDVSVNSVVSTFPMGSTVLDQQLGCLWQKDHLLSVSLSGYINYLDRNNPSKPLHVIKGHSKSIQCLTVHKNGGKSYIYSGSHDGHINYWDSETGENDSFAGKGHTNQVSRMTVDESGQLISCSMDDTVRYTSLMLRDYSGQGVVKLDVQPKCVAVGPGGYAVVVCIGQIVLLKDQRKCFSIDNPGYEPEVVAVHPGGDTVAIGGVDGNVRLYSILGTTLKDEGKLLEAKGPVTDVAYSHDGAFLAVCDASKVVTVFSVADGYSENNVFYGHHAKIVCLAWSPDNEHFASGGMDMMVYVWTLSDPETRVKIQDAHRLHHVSSLAWLDEHTLVTTSHDASVKEWTITY
RUO
WDR1
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AIP1|NORI-1
WDR1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit