Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDR16 Full-length ORF (AAH25392.1, 1 a.a. - 620 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00146845P01L

 View more versions of this product

Catalog No. 89-967-273


Add to cart

Description

Description

WD repeat-containing proteins, such as WDR16, play crucial roles in a wide range of physiologic functions, including signal transduction, RNA processing, remodeling the cytoskeleton, regulation of vesicular traffic, and cell division (Silva et al., 2005 [PubMed 15967112]).[supplied by OMIM]

Sequence: MDNKISPEAQVAELELDAVIGFNGHVPTGLKCHPDQEHMIYPLGCTVLIQAINTKEQNFLQGHGNNVSCLAISRSGEYIASGQVTFMGFKVDIILWDYKNRELLARLSLHKGKIEALAFSPNDLYLVSLGGPDDGSVVVWSIAKRDAICGSPAAGLNVGNATNVIFSRCRDEMFMTAGNGTIRVWELDLPNRKIWPTESQTGQLKRIVMSIGVDDDDSFFYLGTTTGDILKMNPRTKLLTDVGPAKDKFSLGVSAIRCLKMGGLLVGSGAGLLVFCKSPGYKPIKKIQLQGGITSITLRGEGHQFLVGTEESHIYRVSFTDFKETLIATCHFDAVKDIVFPFGTAELFATCAKKDIRVWHTSSNRELLRITVPNMTCHGIDFMRDGKSIISAWNDGKIRAFAPETGRLMYVINNAHRIGVTAIATTSDCKRVISGGGEGEVRVWQIGCQTQKLEEALKEHKSSVSCIRVKRNNEECVTASTDGTCIIWDLVRLRRNQMILANTLFQCVCYHPEEFQIITSGTDRKIAYWEVFDGTVIRELEGSLSGSINGMDITQEGVHFVTGGNDHLVKVWDYNEGEVTHVGVGHSGNITRIRISPGNQYIVSVSADGAILRWKYPYTS
Specifications

Specifications

AAH25392.1
Liquid
146845
WDR16 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDNKISPEAQVAELELDAVIGFNGHVPTGLKCHPDQEHMIYPLGCTVLIQAINTKEQNFLQGHGNNVSCLAISRSGEYIASGQVTFMGFKVDIILWDYKNRELLARLSLHKGKIEALAFSPNDLYLVSLGGPDDGSVVVWSIAKRDAICGSPAAGLNVGNATNVIFSRCRDEMFMTAGNGTIRVWELDLPNRKIWPTESQTGQLKRIVMSIGVDDDDSFFYLGTTTGDILKMNPRTKLLTDVGPAKDKFSLGVSAIRCLKMGGLLVGSGAGLLVFCKSPGYKPIKKIQLQGGITSITLRGEGHQFLVGTEESHIYRVSFTDFKETLIATCHFDAVKDIVFPFGTAELFATCAKKDIRVWHTSSNRELLRITVPNMTCHGIDFMRDGKSIISAWNDGKIRAFAPETGRLMYVINNAHRIGVTAIATTSDCKRVISGGGEGEVRVWQIGCQTQKLEEALKEHKSSVSCIRVKRNNEECVTASTDGTCIIWDLVRLRRNQMILANTLFQCVCYHPEEFQIITSGTDRKIAYWEVFDGTVIRELEGSLSGSINGMDITQEGVHFVTGGNDHLVKVWDYNEGEVTHVGVGHSGNITRIRISPGNQYIVSVSADGAILRWKYPYTS
RUO
WDR16
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
94.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37528|WDRPUH
WDR16
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit