Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDR19 Full-length ORF (AAH32578.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057728P01L

 View more versions of this product

Catalog No. 89-967-280

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains six WD repeats, a clathrin heavy-chain repeat, and three transmembrane domains. This gene is conserved from C. elegans to human. It may participate in androgen-regulated signaling mechanisms or in the vesicular trafficking of androgen-regulated secretory processes. Alternatively spliced transcript variants encoding distinct isoforms have been reported but the full-length nature of one of these variants has not been defined. [provided by RefSeq]

Sequence: MKRIFSLLEKTWLGAPIQFAWQKTSGNYLAVTGADYIVKIFDRHGQKRSEINLPGNCVAMDWDKDGDVLAVIAEKSSCIYLWDANTNKTSQLDNGMRDQMSFLLWSKVGSFLAVGTVKGNLLIYNHQTSRKIPVLGKHTKRITCGCWNAENLLALGGEDKMITVSNQEGDTIRQTQVRSEPSNMQFFLMKMDDRTSAAESMISVVLGKKTLFFLNLNEPDNPADLEFQQDFGNIVCYNWYGDGRIMIGFSCGHFVVISTHTGELGQEIFQARNHKDNLTSIAVSQTLNKVATCGDNCIKIQDLVDLKDMYVILNLDEENKGLGTLSWTDDGQLLALSTQRGSLHVFLTKLPILGDACSTRIAYLTSLLEVTVANPVEGELPITVSVDVEPNFVAVGLYHLAVGMNNRAWFYVLGENAVKKIERYGVSGNSSQYLPSF
Specifications

Specifications

AAH32578.1
Liquid
57728
WDR19 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKRIFSLLEKTWLGAPIQFAWQKTSGNYLAVTGADYIVKIFDRHGQKRSEINLPGNCVAMDWDKDGDVLAVIAEKSSCIYLWDANTNKTSQLDNGMRDQMSFLLWSKVGSFLAVGTVKGNLLIYNHQTSRKIPVLGKHTKRITCGCWNAENLLALGGEDKMITVSNQEGDTIRQTQVRSEPSNMQFFLMKMDDRTSAAESMISVVLGKKTLFFLNLNEPDNPADLEFQQDFGNIVCYNWYGDGRIMIGFSCGHFVVISTHTGELGQEIFQARNHKDNLTSIAVSQTLNKVATCGDNCIKIQDLVDLKDMYVILNLDEENKGLGTLSWTDDGQLLALSTQRGSLHVFLTKLPILGDACSTRIAYLTSLLEVTVANPVEGELPITVSVDVEPNFVAVGLYHLAVGMNNRAWFYVLGENAVKKIERYGVSGNSSQYLPSF
RUO
WDR19
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ23127|KIAA1638|ORF26|PWDMP
WDR19
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit