Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDR5 Partial ORF (NP_060058, 16 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011091Q01L

 View more versions of this product

Catalog No. 89-967-311


Add to cart

Description

Description

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 7 WD repeats. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq]

Sequence: QPTPSSSATQSKPTPVKPNYALKFTLAGHTKAVSSVKFSPNGEWLASSSADKLIKIWGAYDGKFEKTISGHKLGISDVAWSSDSNLLVSASDDKTLKIWDVSSGK
Specifications

Specifications

NP_060058
Liquid
11091
Human WDR5 Partial ORF (NP_060058, 16 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
25μg
RUO
WDR5
wheat germ expression system
Yes
AP, Array, ELISA, WB-Re
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
BIG-3, SWD3
WDR5
Wheat Germ (in vitro)
GST Tag
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit