Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human WDR5B Full-length ORF (NP_061942.2, 1 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00054554P01L
Description
This intronless gene encodes a protein containing several WD40 repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, including a trp-asp at the C-terminal end. The encoded protein may mediate protein-protein interactions. [provided by RefSeq]
Sequence: MATKESRDAKAQLALSSSANQSKEVPENPNYALKCTLVGHTEAVSSVKFSPNGEWLASSSADRLIIIWGAYDGKYEKTLYGHNLEISDVAWSSDSSRLVSASDDKTLKLWDVRSGKCLKTLKGHSNYVFCCNFNPPSNLIISGSFDETVKIWEVKTGKCLKTLSAHSDPVSAVHFNCSGSLIVSGSYDGLCRIWDAASGQCLKTLVDDDNPPVSFVKFSPNGKYILTATLDNTLKLWDYSRGRCLKTYTGHKNEKYCIFANFSVTGGKWIVSGSEDNLVYIWNLQTKEIVQKLQGHTDVVISAACHPTENLIASAALENDKTIKLWMSNHSpecifications
NP_061942.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11287/MGC49879 | |
WDR5B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
54554 | |
WDR5B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATKESRDAKAQLALSSSANQSKEVPENPNYALKCTLVGHTEAVSSVKFSPNGEWLASSSADRLIIIWGAYDGKYEKTLYGHNLEISDVAWSSDSSRLVSASDDKTLKLWDVRSGKCLKTLKGHSNYVFCCNFNPPSNLIISGSFDETVKIWEVKTGKCLKTLSAHSDPVSAVHFNCSGSLIVSGSYDGLCRIWDAASGQCLKTLVDDDNPPVSFVKFSPNGKYILTATLDNTLKLWDYSRGRCLKTYTGHKNEKYCIFANFSVTGGKWIVSGSEDNLVYIWNLQTKEIVQKLQGHTDVVISAACHPTENLIASAALENDKTIKLWMSNH | |
RUO | |
WDR5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction