Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WDR73 Full-length ORF (NP_116245.2, 1 a.a. - 378 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$434.00 - $788.00

Specifications

Accession Number NP_116245.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84942
Molecular Weight (g/mol) 68.1kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-967-344
View Documents
Abnova™
H00084942P01L
25 ug
Each for $788.00
Add to Cart
 
89-967-343
View Documents
Abnova™
H00084942P01S
10 ug
Each for $434.00
Add to Cart
 
Description

Description

Sequence: MDPGDDWLVESLRLYQDFYAFDLSGATRVLEWIDDKGVFVAGYESLKKNEILHLKLPLRLSVKENKGLFPERDFKVRHGGFSDRSIFDLKHVPHTRLLVTSGLPGCYLQVWQVAEDSDVIKAVSTIAVHEKEESLWPRVAVFSTLAPGVLHGARLRSLQVVDLESRKTTYTSDVSDSEELSSLQVLDADTFAFCCASGRLGLVDTRQKWAPLENRSPGPGSGGERWCAEVGSWGQGPGPSIASLGSDGRLCLLDPRDLCHPVSSVQCPVSVPSPDPELLRVTWAPGLKNCLAISGFDGTVQVYDATSWDGTRSQDGTRSQVEPLFTHRGHIFLDGNGMDPAPLVTTHTWHPCRPRTLLSATNDASLHVWDWVDLCAPR
Specifications

Specifications

NP_116245.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.1kDa
Glutathione Sepharose 4 Fast Flow
MDPGDDWLVESLRLYQDFYAFDLSGATRVLEWIDDKGVFVAGYESLKKNEILHLKLPLRLSVKENKGLFPERDFKVRHGGFSDRSIFDLKHVPHTRLLVTSGLPGCYLQVWQVAEDSDVIKAVSTIAVHEKEESLWPRVAVFSTLAPGVLHGARLRSLQVVDLESRKTTYTSDVSDSEELSSLQVLDADTFAFCCASGRLGLVDTRQKWAPLENRSPGPGSGGERWCAEVGSWGQGPGPSIASLGSDGRLCLLDPRDLCHPVSSVQCPVSVPSPDPELLRVTWAPGLKNCLAISGFDGTVQVYDATSWDGTRSQDGTRSQVEPLFTHRGHIFLDGNGMDPAPLVTTHTWHPCRPRTLLSATNDASLHVWDWVDLCAPR
RUO
WDR73
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84942
WDR73 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14888/HSPC264
WDR73
Recombinant
wheat germ expression system
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit