Learn More
Abnova™ Human WIPI1 Full-length ORF (NP_060453.2, 1 a.a. - 446 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00055062P01S
Description
WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI1, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids (Proikas-Cezanne et al., 2004 [PubMed 15602573]).[supplied by OMIM]
Sequence: MEAEAADAPPGGVESALSCFSFNQDCTSLAIGTKAGYKLFSLSSVEQLDQVHGSNEIPDVYIVERLFSSSLVVVVSHTKPRQMNVYHFKKGTEICNYSYSSNILSIRLNRQRLLVCLEESIYIHNIKDMKLLKTLLDIPANPTGLCALSINHSNSYLAYPGSLTSGEIVLYDGNSLKTVCTIAAHEGTLAAITFNASGSKLASASEKGTVIRVFSVPDGQKLYEFRRGMKRYVTISSLVFSMDSQFLCASSNTETVHIFKLEQVTNSRPEEPSTWSGYMGKMFMAATNYLPTQVSDMMHQDRAFATARLNFSGQRNICTLSTIQKLPRLLVASSSGHLYMYNLDPQDGGECVLIKTHSLLGSGTTEENKENDLRPSLPQSYAATVARPSASSASTVPGYSEDGGALRGEVIPEHEFATGPVCLDDENEFPPIILCRGNQKGKTKQSSpecifications
NP_060453.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATG18/FLJ10055/WIPI49 | |
WIPI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55062 | |
WIPI1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEAEAADAPPGGVESALSCFSFNQDCTSLAIGTKAGYKLFSLSSVEQLDQVHGSNEIPDVYIVERLFSSSLVVVVSHTKPRQMNVYHFKKGTEICNYSYSSNILSIRLNRQRLLVCLEESIYIHNIKDMKLLKTLLDIPANPTGLCALSINHSNSYLAYPGSLTSGEIVLYDGNSLKTVCTIAAHEGTLAAITFNASGSKLASASEKGTVIRVFSVPDGQKLYEFRRGMKRYVTISSLVFSMDSQFLCASSNTETVHIFKLEQVTNSRPEEPSTWSGYMGKMFMAATNYLPTQVSDMMHQDRAFATARLNFSGQRNICTLSTIQKLPRLLVASSSGHLYMYNLDPQDGGECVLIKTHSLLGSGTTEENKENDLRPSLPQSYAATVARPSASSASTVPGYSEDGGALRGEVIPEHEFATGPVCLDDENEFPPIILCRGNQKGKTKQS | |
RUO | |
WIPI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.