Learn More
Raybiotech IncSupplier Diversity Partner Human XIAP ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHXIAP1
For cell culture supernatants, plasma, and serum samples. Sensitivity: 250 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair can recognize human and mouse XIAP.
Catalog No. 50-194-9862