Learn More
Abnova™ Human YES1 (NM_005433, 1 a.a. - 543 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4797
Catalog No. 89-967-597
Add to cart
Description
Description
This gene is the cellular homolog of the Yamaguchi sarcoma virus oncogene. The encoded protein has tyrosine kinase activity and belongs to the src family of proteins. This gene lies in close proximity to thymidylate synthase gene on chromosome 18, and a corresponding pseudogene has been found on chromosome 22. [provided by RefSeq]
Specifications
Specifications
Insect cell (Sf9) expression system
0.116 ug/uL
Liquid
7525
YES1 (Human) Recombinant Protein
One-step affinity purification using GSH-agarose
100 ug
RUO
YES1
29pmol/μg x min
Yes
NM_005433
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
90.997kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
HsT441/P61-YES/Yes/c-yes
YES1
Insect
GST-His